2,5-Dimethyl-2,4-hexadiene ≥96%

Supplier: BeanTown Chemical

Synonyms: 2,5-Dimethylhexa-2,4-diene

131935-10G 131935-250G 131935-50G
BT131935-10GEA 33.59 USD
BT131935-10G BT131935-250G BT131935-50G
2,5-Dimethyl-2,4-hexadiene ≥96%
2,5-Dimethyl-2,4-hexadiene

CAS: 764-13-6; EC No: 212-115-5; MDL No: MFCD00008904 UN No: UN3295; Haz Class: 3; Packing Group: III Liquid; Linear Formula: (CH3)2C=CHCH=C(CH3)2; Molecular Formula: C8H14; MW: 110.20 Melting Point: 11-14°; Boiling Point: 132-134°; Flash point: 29°C (84°F) Density (g/mL): 0.773 ; Refractive Index: 1.476


  • Refractive index: 1.476
  • Liquid

Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 132…134 °C
Melting Pt: 11…14 °C
Density: 0.773 g/mL
Flash Pt: 29 °C (84 °F)
MDL Number: MFCD00008904
CAS Number: 764-13-6
EINECS: 212-115-5
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR