Tetrapropylammonium hydroxide 40% (w/w) in water
Supplier: BeanTown Chemical
CAS: 4499-86-9; EC No: 224-800-6; MDL No: MFCD00009360; RTECS: BS8400000
UN No: UN3267; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: (CH3CH2CH2)4NOH ; MW: 203.37
Boiling Point: 102°
Density (g/mL): 0.99; Refractive Index: 1.372
Air Sensitive
- RTECS: BS8400000
- Refractive index: 1.372
- Air Sensitive
- Liquid
Formula:
C₁₂H₂₉NO Boiling Pt: 102 °C Density: 0.99 g/mL |
MDL Number:
MFCD00009360 CAS Number: 4499-86-9 EINECS: 224-800-6 UN: 3267 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...