Lead(II) zirconate ≥99.7% (trace metals basis excluding Hf)
Supplier: BeanTown Chemical
131885-100G
131885-500G
BT131885-100GEA
204
USD
BT131885-100G
BT131885-500G
Lead(II) zirconate ≥99.7% (trace metals basis excluding Hf)
Lead(II) zirconate
CAS: 12060-01-4; EC No: 235-039-4; MDL No: MFCD00016275
UN No: UN3077; Haz Class: 9; Packing Group: III
-325 Mesh Powder; Linear Formula: PbZrO3; MW: 346.42
- -325 Mesh Powder
Formula:
O₃PbZr MW: 346.421 g/mol |
MDL Number:
MFCD00016275 CAS Number: 12060-01-4 EINECS: 235-039-4 ADR: 9 III |
Specification Test Results
Assay | 99.7% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...