Diphenylphosphinic chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
Diphenylphosphinyl chloride
131880-100G
131880-25G
131880-5G
BT131880-100GEA
400
USD
BT131880-100G
BT131880-25G
BT131880-5G
Diphenylphosphinic chloride ≥97%
Diphenylphosphinic chloride
CAS: 1499-21-4; EC No: 216-107-2; MDL No: MFCD00002077
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: (C6H5)2P(O)Cl; Molecular Formula: C12H10ClOP; MW: 236.64
Boiling Point: 222°/16 mmHg; Flash point: 26°C (79°F)
Density (g/mL): 1.24; Refractive Index: 1.609
Moisture Sensitive
- Refractive index: 1.609
- Moisture Sensitive
- Liquid
Formula:
C₁₂H₁₀ClOP MW: 236.63 g/mol Boiling Pt: 222 °C/16 mmHg Density: 1.24 g/mL Flash Pt: 26 °C (79 °F) |
MDL Number:
MFCD00002077 CAS Number: 1499-21-4 EINECS: 216-107-2 UN: 2920 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...