1-Fluoro-2-nitrobenzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
o-Fluoronitrobenzene, 2-Fluoronitrobenzene
131830-1KG
131830-250G
131830-50G
BT131830-1KGEA
556
USD
BT131830-1KG
BT131830-250G
BT131830-50G
1-Fluoro-2-nitrobenzene ≥99%
1-Fluoro-2-nitrobenzene
CAS: 1493-27-2; EC No: 216-088-0; MDL No: MFCD00007048
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H4FNO2 ; MW: 141.10
Melting Point: -9°; Boiling Point: 116°/22 mmHg; Flash point: 94°C (201°F)
Density (g/mL): 1.338; Refractive Index: 1.532
- Refractive index: 1.532
- Liquid
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Boiling Pt: 116 °C/22 mmHg Melting Pt: –9 °C Density: 1.338 g/mL Flash Pt: 94 °C (201 °F) |
MDL Number:
MFCD00007048 CAS Number: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...