2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite ≥95%
Supplier: BeanTown Chemical
131810-1G
131810-5G
BT131810-1GEA
135.54
USD
BT131810-1G
BT131810-5G
2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite ≥95%
2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite
CAS: 89992-70-1; MDL No: MFCD00011544
UN No: UN2845; Haz Class: 4.2; Packing Group: I
Liquid; Molecular Formula: C9H18ClN2OP; MW: 236.68
Flash point: 110°C (230°F)
Density (g/mL): 1.061
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₉H₁₈ClN₂OP MW: 236.68 g/mol Density: 1.061 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00011544 CAS Number: 89992-70-1 ADR: 4.2,I |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...