4-Bromo-o-xylene ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Bromo-1,2-dimethylbenzene
131795-100G
131795-25G
131795-5G
BT131795-100GEA
301
USD
BT131795-100G
BT131795-25G
BT131795-5G
4-Bromo-o-xylene ≥97%
4-Bromo-o-xylene
CAS: 583-71-1; EC No: 209-518-3; MDL No: MFCD00000086; RTECS: CY9020400
UN No: UN1701; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C8H9Br ; MW: 185.06
Boiling Point: 215°; Flash point: 80°C (176°F)
Density (g/mL): 1.37; Refractive Index: 1.555
- RTECS: CY9020400
- Refractive index: 1.555
- Liquid
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 215 °C Density: 1.37 g/mL Flash Pt: 80 °C (176 °F) |
MDL Number:
MFCD00000071 CAS Number: 583-71-1 EINECS: 209-518-3 UN: 1701 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...