(R)-(+)-1-(1-Naphthyl)ethylamine ≥98%, ee 98% ee
Supplier: BeanTown Chemical
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine
131785-1G
131785-25G
131785-5G
BT131785-1GEA
62.97
USD
BT131785-1G
BT131785-25G
BT131785-5G
(R)-(+)-1-(1-Naphthyl)ethylamine ≥98%, ee 98% ee
(R)-(+)-1-(1-Naphthyl)ethylamine
CAS: 3886-70-2; EC No: 223-425-5; MDL No: MFCD00064114
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III
Liquid; Molecular Formula: C12H13N; MW: 171.24
Boiling Point: 153°/11 mmHg; Flash point:
- Optical rotation: [α]20/D +55°, c = 2 in ethanol
- Refractive index: 1.623
- Air Sensitive
- Liquid
Formula:
C₁₂H₁₃N MW: 171.24 g/mol Boiling Pt: 153 °C/11 mmHg Density: 1.067 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00064114 CAS Number: 3886-70-2 EINECS: 223-425-5 ADR: 8 (6.1),III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...