Trimethyl orthoacetate ≥98%

Supplier: BeanTown Chemical

131765-250ML 131765-50ML
BT131765-250MLEA 100 USD
BT131765-250ML BT131765-50ML
Trimethyl orthoacetate ≥98%
Trimethyl orthoacetate

CAS: 1445-45-0; EC No: 215-892-9; MDL No: MFCD00008477 UN No: UN3272; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3C(OCH3)3; Molecular Formula: C5H12O3; MW: 120.15 Boiling Point: 107-109°; Flash point: 15°C (59°F) Density (g/mL): 0.944; Refractive Index: 1.388 Moisture Sensitive


  • Refractive index: 1.388
  • Moisture Sensitive
  • Liquid

Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 107…109 °C
Density: 0.944 g/mL
Flash Pt: 15 °C (59 °F)
MDL Number: MFCD00008477
CAS Number: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR