Potassium hexafluorophosphate ≥95%

Supplier: BeanTown Chemical

131750-1KG 131750-250G
BT131750-1KGEA 182.57 USD
BT131750-1KG BT131750-250G
Potassium hexafluorophosphate ≥95%
Potassium hexafluorophosphate

CAS: 17084-13-8; EC No: 241-143-0; MDL No: MFCD00011412 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: KPF6; MW: 184.07 Melting Point: 575° Density (g/mL): 2.75 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: F₆KP
MW: 184.06 g/mol
Melting Pt: 575
Density: 2.75 g/mL
MDL Number: MFCD00011412
CAS Number: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR