Nickel(II) tetrafluoroborate hexahydrate

Supplier: BeanTown Chemical

131745-100G 131745-500G
BT131745-100GEA 123.5 USD
BT131745-100G BT131745-500G
Nickel(II) tetrafluoroborate hexahydrate
Nickel(II) tetrafluoroborate hexahydrate

CAS: 15684-36-3; EC No: 238-753-4; MDL No: MFCD00016249; RTECS: QR6650000 UN No: UN3077; Haz Class: 9; Packing Group: III Lump; Linear Formula: Ni(BF4)2·6H2O; MW: 340.40 Density (g/mL): 1.5 Hygroscopic


  • RTECS: QR6650000
  • Hygroscopic
  • Lump

Formula: B₂F₈Ni·6H₂O
MW: 340.394 g/mol
Density: 1.5 g/mL
MDL Number: MFCD00150259
CAS Number: 15684-36-3
EINECS: 238-753-4
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR