Potassium osmate(VI) dihydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Potassium osmium(VI) oxide dihydrate, Potassium dioxidodioxoosmium dihydrate, Dipotassium osmate dihydrate
131735-1G
131735-250MG
BT131735-1GEA
233
USD
BT131735-1G
BT131735-250MG
Potassium osmate(VI) dihydrate ≥99%
Potassium osmate(VI) dihydrate
CAS: 10022-66-9; EC No: 243-247-1; MDL No: MFCD00149919
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: K2OsO4·2H2O; MW: 368.43
Hygroscopic
- Hygroscopic
- Powder
Formula:
K₂O₄Os·2H₂O MW: 368.45 g/mol |
MDL Number:
MFCD00149919 CAS Number: 10022-66-9 EINECS: 243-247-1 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...