Benzyl chloride ≥99% stabilized
Supplier: BeanTown Chemical
Synonyms:
(Chloromethyl)benzene, Chlorophenylmethane, α-Chlorotoluene
131720-10KG
131720-2.5KG
131720-500G
BT131720-10KGEA
278
USD
BT131720-10KG
BT131720-2.5KG
BT131720-500G
Benzyl chloride ≥99% stabilized
Benzyl chloride
CAS: 100-44-7; EC No: 202-853-6; MDL No: MFCD00000889; RTECS: XS8925000
UN No: UN1738; Haz Class: 6.1 (8); Packing Group: II
Liquid; Linear Formula: C6H5CH2Cl; Molecular Formula: C7H7Cl; MW: 126.58
Melting Point: -43°; Boiling Point: 177-181°; Flash point: 60°C (140°F)
Density (g/mL): 1.1; Refractive Index: 1.538
- RTECS: XS8925000
- Refractive index: 1.538
- Liquid
Formula:
(C₆H₅)CH₂Cl MW: 126.59 g/mol Boiling Pt: 177…181 °C Melting Pt: –43 °C Density: 1.1 g/mL Flash Pt: 60 °C (140 °F) |
MDL Number:
MFCD00000889 CAS Number: 100-44-7 EINECS: 202-853-6 UN: 1738 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...