Diphenylamine ACS

Supplier: BeanTown Chemical

131690-100G 131690-25G 131690-500G
BT131690-100GEA 69.88 USD
BT131690-100G BT131690-25G BT131690-500G
Diphenylamine ACS
Diphenylamine

CAS: 122-39-4; EC No: 204-539-4; MDL No: MFCD00003014; RTECS: JJ7800000 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Linear Formula: (C6H5)2NH; Molecular Formula: C12H11N; MW: 169.23 Melting Point: 50-53°; Boiling Point: 302°; Flash point: 153°C (307°F) Air Sensitive, Light Sensitive


  • RTECS: JJ7800000
  • Air Sensitive, Light Sensitive
  • Solid

Formula: (C₆H₅)₂NH
Boiling Pt: 302 °C
Melting Pt: 50…53 °C
Flash Pt: 153 °C (307 °F)
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR