1,1,3,3-Tetramethylguanidine ≥99%
Supplier: BeanTown Chemical
CAS: 80-70-6; EC No: 201-302-7; MDL No: MFCD00008323
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: C5H13N3; MW: 115.18
Boiling Point: 52-54°/11 mmHg; Flash point: 60°C (140°F)
Density (g/mL): 0.918; Refractive Index: 1.407
Air Sensitive
- Refractive index: 1.407
- Air Sensitive
- Liquid
Formula:
C₅H₁₃N₃ Boiling Pt: 52…54 °C/11 mmHg Density: 0.918 g/mL Flash Pt: 60 °C (140 °F) |
MDL Number:
MFCD00008323 CAS Number: 80-70-6 EINECS: 201-302-7 UN: 2920 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...