Chlorodimethyloctadecylsilane ≥95%

Supplier: BeanTown Chemical

131610-100G 131610-25G 131610-5G
BT131610-100GEA 298 USD
BT131610-100G BT131610-25G BT131610-5G
Chlorodimethyloctadecylsilane ≥95%
Chlorodimethyloctadecylsilane

CAS: 18643-08-8; EC No: 242-472-2; MDL No: MFCD00000504 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Linear Formula: CH3(CH2)17Si(CH3)2Cl; Molecular Formula: C20H43ClSi ; MW: 347.09 Melting Point: 28-30°; Boiling Point: 145-155°/0.005 mmHg; Flash point: 200°C (392°F) Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₂₀H₄₃ClSi
MW: 347.10 g/mol
Boiling Pt: 145…155 °C/0.005 mmHg
Melting Pt: 28…30 °C
Flash Pt: 200 °C (392 °F)
MDL Number: MFCD00000504
CAS Number: 18643-08-8
EINECS: 242-472-2
ADR: 8,II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR