Nickel (II) sulfate hexahydrate ≥98.0% ACS

Supplier: BeanTown Chemical

131595-100G 131595-2.5KG 131595-500G
BT131595-100GEA 33.29 USD
BT131595-100G BT131595-2.5KG BT131595-500G
Nickel (II) sulfate hexahydrate ≥98.0% ACS
Nickel(II) sulphate hexahydrate

CAS: 10101-97-0; EC No: 232-104-9; MDL No: MFCD00149813; RTECS: QR9600000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: NiSO4·6H2O; MW: 262.86 Density (g/mL): 2.070


  • RTECS: QR9600000
  • Crystalline/Powder

Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Density: 2.070 g/mL
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98.0% min.
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.001%
Nitrogen compounds (as N) ≤0.002%
Calcium (Ca) ≤0.005%
Cobalt (Co) ≤0.002%
Copper (Cu) ≤0.005%
Iron (Fe) ≤0.001%
Magnesium (Mg) ≤0.005%
Manganese (Mn) ≤0.002%
Potassium (K) ≤0.01%
Sodium (Na) ≤0.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR