Copper(II) nitrate hydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
CAS: 13778-31-9; EC No: 221-838-5; MDL No: MFCD00149669; RTECS: GL7875000
UN No: UN3085; Haz Class: 5.1 (8); Packing Group: III
Crystalline Aggregates; Linear Formula: Cu(NO3)2·xH2O; MW: 187.55 (anhydrous)
Density (g/mL): 2.32
Hygroscopic
- RTECS: GL7875000
- Hygroscopic
- Crystalline Aggregates
Formula:
CuN₂O₆·H₂O Density: 2.32 g/mL |
MDL Number:
MFCD00149669 CAS Number: 13778-31-9 EINECS: 221-838-5 UN: 3085 ADR: 5.1,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...