Guanosine hydrate ≥98%

Supplier: BeanTown Chemical

131575-100G 131575-25G
BT131575-100GEA 130 USD
BT131575-100G BT131575-25G
Guanosine hydrate ≥98%
Guanosine hydrate

CAS: 118-00-3; EC No: 204-227-8; MDL No: MFCD00010182; RTECS: MF8750000 UN No: UN2811 ; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C10H13N5O5; MW: 283.24 Melting Point: 250° (decomposes)


  • RTECS: MF8750000
  • Crystalline

Formula: C₁₀H₁₃N₅O₅·H₂O
Melting Pt: 250 °C (dec.)
CAS Number: 118-00-3
EINECS: 204-227-8
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR