5-Fluorouracil ≥99%

Supplier: BeanTown Chemical

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

131560-100G 131560-25G 131560-5G
BT131560-100GEA 1031 USD
BT131560-100G BT131560-25G BT131560-5G
5-Fluorouracil ≥99%
5-Fluorouracil

CAS: 51-21-8; EC No: 200-085-6; MDL No: MFCD00006018; RTECS: YR0350000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C4H3FN2O2; MW: 130.08 Melting Point: 282-286° (decomposes) Light Sensitive


  • RTECS: YR0350000
  • Light Sensitive
  • Powder

Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347
Melting Pt: 282…286 °C (dec.)
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,04208

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR