N-Phenylmaleimide ≥98%
Supplier: BeanTown Chemical
131555-100G
131555-25G
131555-500G
BT131555-100GEA
156
USD
BT131555-100G
BT131555-25G
BT131555-500G
N-Phenylmaleimide ≥98%
N-Phenylmaleimide
CAS: 941-69-5; EC No: 213-382-0; MDL No: MFCD00005502; RTECS: ON5950000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Flakes; Molecular Formula: C10H7NO2; MW: 173.17
Melting Point: 85-87°; Boiling Point: 162-163°/12 mmHg; Flash point: 152°C (306°F)
Density (g/mL): 1.18
Moisture Sensitive
- RTECS: ON5950000
- Moisture Sensitive
- Flakes
Formula:
C₁₀H₇NO₂ Boiling Pt: 162…163 °C/12 mmHg Melting Pt: 85…87 °C Density: 1.18 g/mL Flash Pt: 152 °C (306 °F) |
MDL Number:
MFCD00005502 CAS Number: 941-69-5 EINECS: 213-382-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...