Zinc oxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Zinci oxidum, Zinc (II) oxide

131545-1KG 131545-250G 131545-5KG
BT131545-1KGEA 91.04 USD
BT131545-1KG BT131545-250G BT131545-5KG
Zinc oxide ≥99.9% (trace metals basis)
Zinc oxide

CAS: 1314-13-2; EC No: 215-222-5; MDL No: MFCD00011300; RTECS: ZH4810000 UN No: UN3077; Haz Class: 9; Packing Group: III -200 Mesh Powder; Linear Formula: ZnO; MW: 81.39 Melting Point: 1975° Density (g/mL): 5.61


  • RTECS: ZH4810000
  • -200 Mesh Powder

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/mL
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR