Antimony(III) chloride ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Antimony trichloride

131490-1KG 131490-250G 131490-50G
BT131490-1KGEA 517 USD
BT131490-1KG BT131490-250G BT131490-50G
Antimony(III) chloride ≥99.0% ACS
Antimony (III) chloride

CAS: 10025-91-9; EC No: 233-047-2; MDL No: MFCD00011212; RTECS: CC4900000 UN No: UN1733; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: SbCl3; MW: 228.12 Melting Point: 73.4° Moisture Sensitive


  • RTECS: CC4900000
  • Moisture Sensitive
  • Crystalline

Formula: SbCl₃
MW: 228.12 g/mol
Melting Pt: 73.4 °C
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR