1-Chloro-3-fluorobenzene ≥99%

Supplier: BeanTown Chemical

131470-100G 131470-25G 131470-500G
BT131470-100GEA 161 USD
BT131470-100G BT131470-25G BT131470-500G
1-Chloro-3-fluorobenzene ≥99%
1-Chloro-3-fluorobenzene

CAS: 625-98-9; EC No: 210-919-0; MDL No: MFCD00000569 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H4ClF; MW: 130.55 Boiling Point: 126-128°; Flash point: 20°C (68°F) Density (g/mL): 1.219; Refractive Index: 1.494


  • Refractive index: 1.494
  • Liquid

Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 126…128 °C
Density: 1.219 g/mL
Flash Pt: 20 °C (68 °F)
MDL Number: MFCD00000569
CAS Number: 625-98-9
EINECS: 210-919-0
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR