Cadmium(II) carbonate ≥99.99%
Supplier: BeanTown Chemical
131400-100G
131400-25G
BT131400-100GEA
269
USD
BT131400-100G
BT131400-25G
Cadmium(II) carbonate ≥99.99%
Cadmium(II) carbonate
CAS: 513-78-0; EC No: 208-168-9; MDL No: MFCD00010918
UN No: UN2570; Haz Class: 6.1; Packing Group: III
Powder/Lump; Linear Formula: CdCO3; MW: 172.42
Melting Point: 500°
Density (g/mL): 4.258
- Powder/Lump
Formula:
CCdO₃ Melting Pt: 500 °C Density: 4.258 g/mL |
MDL Number:
MFCD00010918 CAS Number: 513-78-0 EINECS: 208-168-9 UN: 2570 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...