Dibenzofuran ≥98%
Supplier: BeanTown Chemical
CAS: 132-64-9; EC No: 205-071-3; MDL No: MFCD00004968; RTECS: HP4430000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: C12H8O; MW: 168.19
Melting Point: 80-82°; Boiling Point: 154-155°/20 mmHg; Flash point: 130°C (266°F)
- RTECS: HP4430000
- Powder
Formula:
C₁₂H₈O Boiling Pt: 154…155 °C/20 mmHg Melting Pt: 80…82 °C Flash Pt: 130 °C (266 °F) |
MDL Number:
MFCD00004968 CAS Number: 132-64-9 EINECS: 205-071-3 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...