3-Amino-N-methylbenzylamine ≥98%

Supplier: BeanTown Chemical

131365-25G 131365-5G
BT131365-25GEA 216 USD
BT131365-25G BT131365-5G
3-Amino-N-methylbenzylamine ≥98%
3-Amino-N-methylbenzylamine

CAS: 18759-96-1; EC No: 414-570-7; MDL No: MFCD00211072 UN No: UN3259; Haz Class: 8; Packing Group: III Solid; Linear Formula: H2NC6H4CH2NHCH3; Molecular Formula: C8H12N2; MW: 136.19


  • Solid

Formula: C₈H₁₂N₂
MDL Number: MFCD00211072
CAS Number: 18759-96-1
EINECS: 414-570-7
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR