α,α'-Dibromo-m-xylene ≥97%
Supplier: BeanTown Chemical
Synonyms:
1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide
131345-10G
131345-50G
BT131345-10GEA
75.22
USD
BT131345-10G
BT131345-50G
α,α'-Dibromo-m-xylene ≥97%
α,α'-Dibromo-m-xylene
CAS: 626-15-3; EC No: 210-931-6; MDL No: MFCD00000178
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline/Powder; Linear Formula: C6H4(CH2Br)2; Molecular Formula: C8H8Br2; MW: 263.96
Melting Point: 74-76°; Boiling Point: 135-140°
- Crystalline/Powder
Formula:
C₈H₈Br₂ MW: 263.96 g/mol Boiling Pt: 135…140 °C Melting Pt: 74…76 °C |
MDL Number:
MFCD00000178 CAS Number: 626-15-3 EINECS: 210-931-6 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...