Copper(II) acetate monohydrate 98.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

131330-100G 131330-2KG 131330-500G
BT131330-100GEA 53.3 USD
BT131330-100G BT131330-2KG BT131330-500G
Copper(II) acetate monohydrate 98.0-102.0% ACS
Copper(II) acetate monohydrate

CAS: 6046-93-1; EC No: 205-553-3; MDL No: MFCD00149570; RTECS: AG3500000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: Cu(OOCCH3)2·H2O; MW: 199.65 Melting Point: 115° Density (g/mL): 1.882


  • RTECS: AG3500000
  • Powder

Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Melting Pt: 115 °C
Density: 1.882 g/mL
MDL Number: MFCD00149570
CAS Number: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98.0-102.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR