4-Chlorophenol ≥99%
Supplier: BeanTown Chemical
Synonyms:
p-Chlorophenol
131305-1KG
131305-250G
131305-50G
BT131305-1KGEA
138
USD
BT131305-1KG
BT131305-250G
BT131305-50G
4-Chlorophenol ≥99%
4-Chlorophenol
CAS: 106-48-9; EC No: 203-402-6; MDL No: MFCD00002318; RTECS: SK2800000
UN No: UN2020; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H5ClO; MW: 128.56
Melting Point: 42-45°; Boiling Point: 220°; Flash point: 121°C (250°F)
Density (g/mL): 1.306
Air Sensitive
- RTECS: SK2800000
- Air Sensitive
- Solid
Formula:
C₆H₅ClO MW: 128.56 g/mol Boiling Pt: 220 °C Melting Pt: 42…45 °C Density: 1.306 g/mL Flash Pt: 121 °C (250 °F) |
MDL Number:
MFCD00002318 CAS Number: 106-48-9 EINECS: 203-402-6 UN: 2020 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...