Lithium dodecyl sulfate ≥99%

Supplier: BeanTown Chemical

131285-10G 131285-50G
BT131285-10GEA 79.41 USD
BT131285-10G BT131285-50G
Lithium dodecyl sulfate ≥99%
Lithium dodecyl sulphate

CAS: 2044-56-6; EC No: 218-058-2; MDL No: MFCD00007467 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Linear Formula: CH3(CH2)11OSO3Li; Molecular Formula: C12H25LiO4S; MW: 272.33


  • Powder

Formula: C₁₂H₂₅LiO₄S
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR