Palladium 5% on carbon Type 87L

Supplier: BeanTown Chemical

131260-10G 131260-50G
BT131260-10GEA 225 USD
BT131260-10G BT131260-50G
Palladium 5% on carbon Type 87L
Palladium

CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Pd; MW: 106.42 Melting Point: 1555°; Boiling Point: 2963°


  • Powder

Formula: Pd
Boiling Pt: 2963 °C
Melting Pt: 1555 °C
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR