2-Fluorobenzoyl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: o-Fluorobenzoyl chloride

131205-100G 131205-25G 131205-500G
BT131205-100GEA 106 USD
BT131205-100G BT131205-25G BT131205-500G
2-Fluorobenzoyl chloride ≥97%
2-Fluorobenzoyl chloride

CAS: 393-52-2; EC No: 206-887-2; MDL No: MFCD00000656; RTECS: DM6640000 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C7H4ClFO ; MW: 158.56 Melting Point: 4°; Boiling Point: 90-92°/15 mmHg; Flash point: 122°C (252°F) Density (g/mL): 1.328; Refractive Index: 1.536 Moisture Sensitive


  • RTECS: DM6640000
  • Refractive index: 1.536
  • Moisture Sensitive
  • Liquid

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 90…92 °C/15 mmHg
Melting Pt: 4 °C
Density: 1.328 g/mL
Flash Pt: 122 °C (252 °F)
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR