Tetrabutyltin ≥95%

Supplier: BeanTown Chemical

Synonyms: Tetrabutylstannane, Tetra-n-butyltin

131180-100G 131180-500G
BT131180-100GEA 56 USD
BT131180-100G BT131180-500G
Tetrabutyltin ≥95%
Tetrabutyltin

CAS: 1461-25-2; EC No: 215-960-8; MDL No: MFCD00009422; RTECS: WH8605000 UN No: UN2788; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: [CH3(CH2)3]4Sn; Molecular Formula: C16H36Sn; MW: 347.17 Melting Point: -97°; Boiling Point: 245-247°; Flash point: 107°C (225°F) Density (g/mL): 1.057 ; Refractive Index: 1.473


  • RTECS: WH8605000
  • Refractive index: 1.473
  • Liquid

Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 145
Melting Pt: -97
Density: 1.057 g/mL
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR