2-Methylbutane 99

Supplier: BeanTown Chemical

Synonyms: iso-Pentane

131095-4X500ML 131095-500ML
BT131095-4X500EA 137.81 USD
BT131095-4X500 BT131095-500ML
2-Methylbutane 99
2-Methylbutane

CAS: 78-78-4; EC No: 201-142-8; MDL No: MFCD00009338; RTECS: EK4430000 UN No: UN1265; Haz Class: 3; Packing Group: I Liquid; Linear Formula: CH3CH2CH(CH3)2; Molecular Formula: C5H12; MW: 72.15 Boiling Point: 30°; Flash point: -51°C (-60°F) Density (g/mL): 0.62; Refractive Index: 1.354


  • RTECS: EK4430000
  • Refractive index: 1.354
  • Liquid

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28
Melting Pt: -160
Density: 0.62 g/mL
Flash Pt: –51 °C (–60 °F)
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR