Selenious acid ≥97%

Supplier: BeanTown Chemical

131085-250G 131085-500G 131085-50G 131085-5G
BT131085-250GEA 228 USD
BT131085-250G BT131085-500G BT131085-50G BT131085-5G
Selenious acid ≥97%
Selenious acid

CAS: 7783-00-8; EC No: 231-974-7; MDL No: MFCD00011331 UN No: UN3290; Haz Class: 6.1 (8); Packing Group: II -4 Mesh Powder; Linear Formula: H2SeO3; MW: 128.97 Melting Point: 70° (decomposes) Density (g/mL): 3.004 Hygroscopic


  • Hygroscopic
  • -4 Mesh Powder

Formula: H₂O₃Se
Melting Pt: 70 °C (dec.)
Density: 3.004 g/mL
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3290
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR