Tributylvinyltin ≥96%

Supplier: BeanTown Chemical

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

131075-1G 131075-5G
BT131075-1GEA 38 USD
BT131075-1G BT131075-5G
Tributylvinyltin ≥96%
Tributylvinyltin

CAS: 7486-35-3; EC No: 231-291-4; MDL No: MFCD00009421 UN No: UN2788; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C14H30Sn; MW: 317.11 Boiling Point: 104-106°; Flash point: 43°C (109°F) Density (g/mL): 1.085; Refractive Index: 1.478


  • Refractive index: 1.478
  • Liquid

Formula: C₁₄H₃₀Sn
MW: 317.1 g/mol
Boiling Pt: 104…106 °C
Density: 1.085 g/mL
Flash Pt: 43 °C (109 °F)
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR