Tin(IV) chloride ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Stannic chloride, Tin tetrachloride
130975-100G
130975-25G
BT130975-100GEA
386.97
USD
BT130975-100G
BT130975-25G
Tin(IV) chloride ≥99.99% (trace metals basis)
Tin(IV) chloride
CAS: 7646-78-8; EC No: 231-588-9; MDL No: MFCD00011242; RTECS: XP8750000
UN No: UN1827; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: SnCl4; MW: 260.52
Melting Point: -33°; Boiling Point: 114°
Density (g/mL): 2.226; Refractive Index: 1.512
Air Sensitive, Moisture Sensitive
- RTECS: XP8750000
- Refractive index: 1.512
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
Cl₄Sn MW: 260.52 g/mol Boiling Pt: 114 °C Melting Pt: –33 °C Density: 2.226 g/mL |
MDL Number:
MFCD00011242 CAS Number: 7646-78-8 EINECS: 231-588-9 UN: 1827 ADR: 8,II |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...