Ammonium cerium(IV) nitrate ≥98.5% ACS

Supplier: BeanTown Chemical

130855-100G 130855-2KG 130855-500G
BT130855-100GEA 77.44 USD
BT130855-100G BT130855-2KG BT130855-500G
Ammonium cerium(IV) nitrate ≥98.5% ACS
Ammonium cerium(IV) nitrate

CAS: 16774-21-3; EC No: 240-827-6; MDL No: MFCD00151121 UN No: UN1477; Haz Class: 5.1; Packing Group: II Granular; Linear Formula: (NH4)2Ce(NO3)6; MW: 548.23


  • Granular

Formula: (NH₄)₂Ce(NO₃)₆
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98.5% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR