tetra-Chloroauric(III) acid trihydrate ≥ 49,5% , 99.99% ACS
Supplier: BeanTown Chemical
Synonyms:
Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate
130830-1G
130830-5G
BT130830-1GEA
155.86
USD
BT130830-1G
BT130830-5G
tetra-Chloroauric(III) acid trihydrate ≥ 49,5% , 99.99% ACS
tetra-Chloroauric(III) acid trihydrate
CAS: 16961-25-4; EC No: 240-948-4; MDL No: MFCD00149904; RTECS: MD5428000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: HAuCl4·3H2O; MW: 339.79 (anhydrous)
Hygroscopic , Light Sensitive
- RTECS: MD5428000
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
HAuCl₄·3H₂O MW: 393.83 g/mol Melting Pt: 30 |
MDL Number:
MFCD00149904 CAS Number: 16961-25-4 EINECS: 240-948-4 UN: 3260 ADR: 8 III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...