Ethyl succinyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 4-chloro-4-oxobutanoate
130820-100G
130820-25G
130820-500G
BT130820-100GEA
193
USD
BT130820-100G
BT130820-25G
BT130820-500G
Ethyl succinyl chloride ≥97%
Ethyl succinyl chloride
CAS: 14794-31-1; EC No: 238-855-9; MDL No: MFCD00000751
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: ClOCCH2CH2CO2CH2CH3; Molecular Formula: C6H9ClO3; MW: 164.59
Boiling Point: 88-90°/11 mmHg; Flash point: 84°C (183°F)
Density (g/mL): 1.155; Refractive Index: 1.437
Moisture Sensitive
- Refractive index: 1.437
- Moisture Sensitive
- Liquid
Formula:
C₆H₉ClO₃ MW: 164.59 g/mol Boiling Pt: 88…90 °C/11 mmHg Density: 1.155 g/mL Flash Pt: 84 °C (183 °F) |
MDL Number:
MFCD00000751 CAS Number: 14794-31-1 EINECS: 238-855-9 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...