3-Acetylpyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone

130795-100G 130795-25G 130795-500G
BT130795-100GEA 134 USD
BT130795-100G BT130795-25G BT130795-500G
3-Acetylpyridine ≥98%
3-Acetylpyridine

CAS: 350-03-8; EC No: 206-496-7; MDL No: MFCD00006396; RTECS: OB5425000 UN No: UN2810; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C7H7NO; MW: 121.14 Melting Point: 11-13°; Boiling Point: 220°; Flash point: 104°C (219°F) Density (g/mL): 1.102; Refractive Index: 1.534


  • RTECS: OB5425000
  • Refractive index: 1.534
  • Liquid

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 220 °C
Melting Pt: 11…13 °C
Density: 1.102 g/mL
Flash Pt: 104 °C (219 °F)
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR