Cesium fluoride ≥99.99%
Supplier: BeanTown Chemical
130785-10G
130785-2G
130785-50G
BT130785-10GEA
134.63
USD
BT130785-10G
BT130785-2G
BT130785-50G
Cesium fluoride ≥99.99%
Cesium fluoride
CAS: 13400-13-0; EC No: 236-487-3; MDL No: MFCD00010960; RTECS: FK9650000
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Crystalline; Molecular Formula: CsF; MW: 151.90
Melting Point: 682°
Density (g/mL): 4.115
Hygroscopic
- RTECS: FK9650000
- Hygroscopic
- Crystalline
Formula:
CsF MW: 151.9 Boiling Pt: 1251 Melting Pt: 680 Density: 4.115 g/mL |
MDL Number:
MFCD00010960 CAS Number: 13400-13-0 EINECS: 236-487-3 UN: 2923 ADR: 8 III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...