Lithium tetrahydroborate ≥95%

Supplier: BeanTown Chemical

Synonyms: Lithium tetrahydridoborate, Lithium borohydride

130780-10G 130780-50G
BT130780-10GEA 247 USD
BT130780-10G BT130780-50G
Lithium tetrahydroborate ≥95%
Lithium tetrahydroborate

CAS: 16949-15-8; EC No: 241-021-7; MDL No: MFCD00011088; RTECS: ED2725000 UN No: UN1413; Haz Class: 4.3; Packing Group: I Powder; Linear Formula: LiBH4; MW: 21.78 Melting Point: 275° (decomposes) Density (g/mL): 0.666 Moisture Sensitive


  • RTECS: ED2725000
  • Moisture Sensitive
  • Powder

Formula: H₄BLi
MW: 21.78 g/mol
Melting Pt: 275 °C (dec.)
Density: 0.666 g/mL
MDL Number: MFCD00011088
CAS Number: 16949-15-8
EINECS: 241-021-7
UN: 1413
ADR: 4.3,I

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR