5-Bromo-2,4-dichloropyrimidine ≥98%

Supplier: BeanTown Chemical

130765-1G 130765-5G
BT130765-1GEA 28 USD
BT130765-1G BT130765-5G
5-Bromo-2,4-dichloropyrimidine ≥98%
5-Bromo-2,4-dichloropyrimidine

CAS: 36082-50-5; MDL No: MFCD00127818 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Solid; Molecular Formula: C4HBrCl2N2; MW: 227.87 Melting Point: 29-30°; Boiling Point: 128°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.781; Refractive Index: 1.603


  • Refractive index: 1.603
  • Solid

Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 128 °C/15 mmHg
Melting Pt: 29…30 °C
Density: 1.781 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR