α-Bromo-3,4-difluorotoluene ≥98%
Supplier: BeanTown Chemical
CAS: 85118-01-0; EC No: 285-653-1; MDL No: MFCD00009889
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C7H5BrF2; MW: 207.02
Flash point: 79°C (174°F)
Density (g/mL): 1.618; Refractive Index: 1.526
- Refractive index: 1.526
- Liquid
Formula:
C₇H₅BrF₂ Density: 1.618 g/mL Flash Pt: 79 °C (174 °F) |
MDL Number:
MFCD00009889 CAS Number: 85118-01-0 EINECS: 285-653-1 UN: 1760 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...