Germanium(IV) iodide ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Tetraiodogermane, Germanium tetraiodide
130750-10G
130750-50G
BT130750-10GEA
249
USD
BT130750-10G
BT130750-50G
Germanium(IV) iodide ≥99.999%
Germanium(IV) iodide
CAS: 13450-95-8; EC No: 236-613-7; MDL No: MFCD00016120
UN No: UN3260; Haz Class: 8; Packing Group: III
-10 Mesh Crystalline; Molecular Formula: GeI4; MW: 580.26
Density (g/mL): 4.32
Moisture Sensitive
- Moisture Sensitive
- -10 Mesh Crystalline
Formula:
GeI₄ Density: 4.32 g/mL |
MDL Number:
MFCD00016120 CAS Number: 13450-95-8 EINECS: 236-613-7 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...