(S)-(-)-Nicotine ≥99%

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine

130705-100G 130705-25G
BT130705-100GEA 208.78 USD
BT130705-100G BT130705-25G
(S)-(-)-Nicotine ≥99%
(S)-(-)-Nicotine

CAS: 54-11-5; EC No: 200-193-3; MDL No: MFCD00006369; RTECS: QS5250000 UN No: UN1654; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C10H14N2; MW: 162.23 Melting Point: -79°; Boiling Point: 245-238°; Flash point: 101°C (213°F) Density (g/mL): 1.01; Refractive Index: 1.526; Optical Rotation: [α]20/D -168±5°, c = 1% in acetone Air Sensitive , Hygroscopic


  • RTECS: QS5250000
  • Optical rotation: [α]20/D -168±5°, c = 1% in acetone
  • Refractive index: 1.526
  • Air Sensitive, Hygroscopic
  • Liquid

Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
Boiling Pt: 245…238 °C
Melting Pt: –79 °C
Density: 1.01 g/mL
Flash Pt: 101 °C (213 °F)
MDL Number: MFCD00006369
CAS Number: 54-11-5
EINECS: 200-193-3
UN: 1654
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR