tert-Butyl nitrite ≥90%, Technical Grade

Supplier: BeanTown Chemical

130690-100G 130690-25G
BT130690-100GEA 66 USD
BT130690-100G BT130690-25G
tert-Butyl nitrite ≥90%, Technical Grade
tert-Butyl nitrite

CAS: 540-80-7; EC No: 208-757-0; MDL No: MFCD00002055; RTECS: RA0802000 UN No: UN2351; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CONO; Molecular Formula: C4H9NO2; MW: 103.12 Boiling Point: 61-63°; Flash point: -23°C (-9°F) Density (g/mL): 0.867; Refractive Index: 1.368


  • RTECS: RA0802000
  • Refractive index: 1.368
  • Liquid

Formula: C₄H₉NO₂
Boiling Pt: 61…63 °C
Density: 0.867 g/mL
Flash Pt: –23 °C (–9 °F)
MDL Number: MFCD00002055
CAS Number: 540-80-7
EINECS: 208-757-0
UN: 2351
ADR: 3,II

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR