trans-Bis(tributylstannyl)ethylene ≥96%

Supplier: BeanTown Chemical

Synonyms: trans-1,2-Bis(tri-n-butylstannyl)ethylene

130665-1G 130665-5G
BT130665-1GEA 125 USD
BT130665-1G BT130665-5G
trans-Bis(tributylstannyl)ethylene ≥96%
trans-Bis(tributylstannyl)ethylene

CAS: 14275-61-7; MDL No: MFCD01631299 UN No: UN2788; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C26H56Sn2; MW: 606.15 Boiling Point: 185°/0.01 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.132; Refractive Index: 1.502 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.502
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₂₆H₅₆Sn₂
MW: 606.15
Boiling Pt: 185 °C/0.01 mmHg
Density: 1.132 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD01631299
CAS Number: 14275-61-7
ADR: 6.1,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR